VITLAB® Griffin Beakers with Molded Graduations, PMP, BrandTech
Supplier: BrandTech
VITLAB®
V61495 V61895 V61695 V61095 V61595 V61195 V61295 V60695 V60995 V60895 V61795 V60795
10218-510PK 50.33 USD
10218-510 10218-532 10218-520 10218-512 10218-522 10218-514 10218-524 10218-516 10218-526 10218-518 10218-528 10218-530
VITLAB® Griffin Beakers with Molded Graduations, PMP, BrandTech
Beakers
High-quality beakers made of glass-transparent polymethylpentene.
  • Glass-clear polymethyl pentene (PMP)
  • Moderate chemical resistance
Beakers feature molded graduations for durability.
Certificaciones: All beakers meet the requirements of ISO 7056, except for 10218-514, 10218-520, and 10218-524, which are variants in addition to ISO 7056.