Barrier Coats, with ESD Stripe, White Knight
Supplier: White Knight
10215-536 10215-527 10215-538 10215-542 10215-534 10215-540 10215-524 10215-530 10215-528 10215-532
10215-529EA 43.09 USD
10215-529 10215-527 10215-538 10215-536 10215-525 10215-534 10215-532 10215-542 10215-531 10215-540
Barrier Coats, with ESD Stripe, White Knight
Workwear
ESD barrier lab coats are made with a fluid-resistant material consisting of 99% polyester and a 1% carbon stripe for electrostatic dissipation (ESD). These barrier coats are ideal for use in medical, healthcare, veterinary, and food processing applications.
  • One breast pocket
  • Two oversized waist pockets
  • Cuffed sleeves
  • Six snap closure
  • Side seam openings for easy access to pants pockets
These coats are extremely durable and will hold up to repeated industrial launderings.
Información para pedidos: Coats in sizes X-small to XX-large are stocked. For sizes 3X-large to 6X-large, extended lead time may be required. Contact your VWR sales representative for more information.