SP Bel-Art Double-Safe PFA Safety Coated, Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art
30599T 30545T 30592T 60171-3911 30591T 30535T 30596T 60171-3711 60171-3811 30515T 30510T 30505T 30593T 30540T
89218-032EA 43.81 USD
89218-032 89218-022 89218-042 89218-036 89218-026 89218-034 89218-046 89218-050 89218-040 10186-274 10186-276 10186-278 89218-038 89218-016
SP Bel-Art Double-Safe PFA Safety Coated, Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
These environmentally safe thermometers are PFA safety coated to keep liquid and glass contained in the event of breakage.
  • Organic liquid fill
  • Non-toxic and biodegradable
  • Lead-free
  • PFA safety coating
  • Yellow back glass
Non-toxic, biodegradable, Enviro-Safe® liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors. They are ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Información para pedidos: Manufactured and tested by our USA quality team, these thermometers are traceable to NIST and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability. Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.