VWR® Individually Calibrated Liquid-In-Glass Thermometer
Supplier: VWR, part of Avantor
VWR®
60205-1311 60170-7611 602010111 602010211 602051311 60205-1111 602050111 60170-7511 60201-0111 60170-7811 602050311 60201-0211 60170-7711 60205-0011 60205-1211 60205-0311 60170-7411 60205-0111
10186-160EA 179 USD
10186-160 CA89095-638 10186-162 CA89095-636 10186-164 CA89095-668 89095-638 10186-156 10186-158 89095-664 89095-632 89095-630 89095-668 CA89095-634 89095-636 CA89095-632 89095-666 89095-634
VWR® Individually Calibrated Liquid-In-Glass Thermometer
Thermometers Analog Thermometers
Individually calibrated, these thermometers feature blue organic liquid fill against lead-free white back glass.
  • Individually serialized
  • Lead-free glass
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
Certificaciones: NIST.
Información para pedidos: Includes individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.