Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Supplier: Omega Bio-Tek
Mag-Bind®
M6246-03 M6246-02 M6246-01 M6246-02 M6246-03 M6246-01
101414-560EA 278.88 USD
CA101414-560 101414-560 CA101414-562 CA76414-002 101414-562 76414-002
Mag-Bind® Viral DNA/RNA 96 Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Mag-Bind® Viral DNA/RNA Kit is designed for the rapid and reliable isolation of viral RNA and viral DNA from whole Blood, serum, plasma, saliva and other body fluids
The Mag-Bind® magnetic beads technology enables purification of high-quality nucleic acids that is free of proteins, nucleases, and other impurities. In addition to easily being adapted with automated systems, this procedure can also be scaled up or down, allowing for the purification from various amounts of starting materials. The purified nucleic acid is ready for direct use in downstream applications, such as amplification or other enzymatic reactions.