BRAND PCR Plates, BrandTech
Supplier: BRANDTECH SCIENTIFIC INC.
781377 781373 781374 781366 781412 781378 781347 781411 781367 781368 781415 781354 781357 781376 781375 781350 781400 781372 781416 781345 781371 781369
89400-714PK 208.19 USD
89400-714 89135-446 89135-456 89135-444 89135-438 89135-458 10141-444 10141-434 80087-150 10141-442 10141-432 10141-440 10141-430 10141-428 10141-426 10141-448 10141-438 89400-702 10141-446 80087-142 10141-436 89400-712
BRAND PCR Plates, BrandTech
Microplates PCR Plates
Designed for applications requiring a high level of accuracy and reproducibility, these 24-well, 48-well, 96-well, and 384-well PCR plates are constructed from high-quality virgin plastics without the addition of lubricants or chemical additives that can inhibit reactions and contaminate samples
  • Ultra-thin walls for optimal heat transfer
  • Cleanroom quality
  • Models to fit popular thermal cyclers
Ultra-thin walled design facilitates constant, rapid, and precise heat transfer. Smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls. Alphanumeric coding and cut corners are embossed and stamped in blue or black. Raised well rims protect against cross-contamination and provide a tight seal in combination with a matching sealing mat. White plastics for qPCR are uniformly colored with titanium dioxide (TiO2) to optimize reflection of the fluorescence signal. Autoclavable at 121°C, per DIN EN 285.
High quality sealing mats and films reduce evaporative losses during PCR process. Roller and cap tool enable easy sealing of films and caps with plates.
These PCR plates are suitable for use with most commercially available thermal cyclers, including low-profile instrumentation.
Certificaciones: DNase-, RNase-, and DNA-free.