VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
Supplier: VWR, part of Avantor
VWR®
131.276.06 131.276.03 131.276.02 131.276.04 131.276.05
10124-198CS 553.81 USD
10124-198 10124-196 10124-114 10124-200 10124-112
VWR® Wide Mouth Volumetric Flasks with PE Stoppers, Class A, Serialized
Flasks Volumetric Flasks
Made from borosilicate 3.3 expansion glass.
  • Borosilicate glass construction
  • Individually serialized
  • Provided with polyethylene stopper
  • Wide-mouthed design
Flasks are serialized and supplied with polyethylene stopper.
Información para pedidos: Flasks come with individual certificate of compliance.