VWR® Color-Coded Autoclavable Instrument Marking Tape
Supplier: VWR, part of Avantor
VWR-IMT-6-C VWR-IMT-3-C VWR-IMT-26-C VWR-IMT-5-C VWR-IMT-37-C VWR-IMT-2-C VWR-IMT-1-C VWR-IMT-4-C VWR-IMT-58-C
10124-154CS 8.41 USD
10124-154 10124-142 10124-152 10124-150 10124-148 10124-158 10124-146 10124-156 10124-144
VWR® Color-Coded Autoclavable Instrument Marking Tape
Tapes
These 9 mm x5m, vinyl, self-stick tapes color code autoclave items.
  • Non-toxic
  • Vinyl
  • Self-stick tapes
Tapes are designed for use on items such as surgical instruments, laboratory glassware, medical waste, and more for easy identification. These tapes attach securely and safely to any clean surface and resist water, oil, and acids.