VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Supplier: VWR, part of Avantor
VWR®
131.236.04A 131.236.02A 131.236.03A 131.236.07A 131.236.05A 131.236.08A 131.236.06A 131.236.01A
10124-088CS 695.73 USD
10124-088 10124-176 10124-174 10124-086 10124-172 10124-094 10124-092 10124-090
VWR® Heavy-Duty Wide Mouth Volumetric Flasks with Glass Stoppers, Amber Glass, Class A, Serialized
Flasks Volumetric Flasks
Heavy-duty wide-mouth volumetric flasks are made to the highest standards to qualify for Class A certification.
  • Borosilicate glass fabrication
  • Flasks are individually serialized
Made from borosilicate 3.3 expansion glass. Flasks are serialized and supplied with glass penny head stopper.
Información para pedidos: Flasks come with individual certificate of compliance.