VWR® Volumetric Pipets, Class A, Serialized, Color-Coded
Supplier: VWR, part of Avantor
VWR®
123.454.04BB 123.454.03BB 123.454.03 123.454.03A 123.454.00A 123.454.07B 123.454.02 123.454.06A 123.454.02C 123.454.09 123.454.02A 123.454.04A 123.454.06C 123.454.03D 123.454.04E
10124-108CS 449.09 USD
10124-108 10124-000 10124-066 10124-110 10124-064 10124-350 10124-360 10124-392 10124-348 10124-006 10124-346 10124-004 10124-002 10124-364 10124-386
VWR® Volumetric Pipets, Class A, Serialized, Color-Coded
Pipets Graduated Pipets
Made from Borosilicate 33 expansion glass, these pipets are calibrated to deliver (TD) at 20 °C as per ASTM E969 standard.
  • Borosilicate glass fabrication
  • Color-coded for easy sorting
  • Individually serialized
Color-coded according to ASTM E1273 for easy sorting and selection. All pipets come with an individual certificate of compliance. A unique three-digit number, along with the batch number, is printed on the pipet for Quality Assurance purposes.