Cryogenic Dot and Strip Labels, VWR, part of Avantor
Supplier: VWR, part of Avantor
VWR®
VWR-C83 VWR-C9 VWR-C22 VWR-C66 VWR-C79 VWR-C41 VWR-C53 VWR-C37 VWR-C8 VWR-C21 VWR-C52 VWR-C32 VWR-C18 VWR-C30 VWR-C7 VWR-C56 VWR-C45 VWR-C70 VWR-C25 VWR-C28 VWR-C69 VWR-C44 VWR-C31 VWR-C24 VWR-C1 VWR-C88 VWR-C82 VWR-C34 VWR-C17 VWR-C64 VWR-C42 VWR-C43 VWR-C14 VWR-C89 VWR-C84 VWR-C26 VWR-C85 VWR-C55 VWR-C36 VWR-C46 VWR-C5 VWR-C63 VWR-C49 VWR-C73 VWR-C72 VWR-C65 VWR-C67 VWR-C95 VWR-C91 VWR-C2 VWR-C12 VWR-C38 VWR-C40 VWR-C96 VWR-C80 VWR-C76 VWR-C27 VWR-C92 VWR-C77 VWR-C15 VWR-C19 VWR-C11 VWR-C54 VWR-C93 VWR-C75 VWR-C10 VWR-C39 VWR-C4 VWR-C16 VWR-C23 VWR-C81 VWR-C90 VWR-C6 VWR-C47 VWR-C71 VWR-C35 VWR-C94 VWR-C51 VWR-C50 VWR-C98 VWR-C33 VWR-C87 VWR-C20 VWR-C29 VWR-C97 VWR-C48 VWR-C3 VWR-C68 VWR-C13
10123-762CS 52.36 USD
10123-762 10123-764 10123-840 10123-766 10123-842 10123-768 10123-844 10123-800 10123-760 89097-578 89097-612 89097-576 89097-610 89097-574 89097-572 10123-836 10123-758 10123-838 89097-616 89097-614 89129-294 10123-772 89129-292 10123-852 10123-774 10123-854 10123-810 89129-290 10123-776 10123-856 10123-812 10123-778 10123-770 89097-600 89097-608 10123-846 10123-802 89097-606 10123-848 10123-804 89097-604 10123-806 89097-602 10123-808 10123-784 89129-282 10123-860 10123-786 89129-280 10123-862 10123-788 10123-820 10123-864 10123-822 10123-866 89097-592 89097-590 89129-288 10123-780 89129-286 10123-782 89129-284 89097-598 89097-596 89097-594 10123-814 10123-858 10123-816 10123-818 10123-872 10123-794 10123-830 10123-874 10123-796 10123-832 10123-798 10123-834 89097-582 89097-580 10123-790 10123-870 10123-792 89097-588 89097-586 89097-584 10123-868 10123-824 10123-826 10123-828
Cryogenic Dot and Strip Labels, VWR, part of Avantor
Labels
VWR® Cryogenic Dot and Strip Labels are ideal for all cryostorage, liquid nitrogen, refrigeration, and ultra-low temperature freezer applications.
  • Labels can be placed in most incubators
Labels feature a unique adhesive that can withstand extreme temperatures from –196...121 °C (–321...250 °F), without the need for lamination.
Información para pedidos: Rolls of labels are supplied on a 44 mm Ø (1³/₄") core. Laser printer label sheets are 21.6W×27.9L cm (8¹/₂×11"). Assorted labels come in blue, green, purple, red, and yellow. Roll of assorted labels contains 1000 labels of each color. Boxes of assorted laser printer labels contain five sheets of each color.