Cleanroom Hoods with ESD Stripe, CritiCore Protective Wear
Supplier: White Knight
10120-388 10120-404 10120-386 10120-394 10120-384 10120-408 10120-378 10120-398 10120-382 10120-392 10120-406 10120-380 10120-402 10120-400 10120-396 10120-390
10120-380EA 26.05 USD
10120-380 10120-392 10120-390 10120-408 10120-406 10120-404 10120-402 10120-388 10120-378 10120-400 10120-386 10120-398 10120-384 10120-396 10120-382 10120-394
Cleanroom Hoods with ESD Stripe, CritiCore Protective Wear
Workwear
White Knight’s reusable, controlled Environment ESD stripe hoods are suitable for a variety of cleanroom applications, including pharmaceutical, biotechnology, and medical device manufacturing.
  • Open face design
  • Made with a high-density polyester calendered fabric that resists fluids and bacterial penetration
  • Hoods feature a tunneled elastic back plus horizontal and vertical adjustment snaps
  • Split-drape design with a front panel that is longer than the back
  • All seams are laser-cut or pre-serged
  • Thread is silicon-free
  • Available in white and navy
  • Sizes range from X-Small to 4X-Large
Designed for cleanroom applications up to and including Class 10 (ISO 4), these hoods also include a one percent carbon stripe for electrostatic dissipation.
Información para pedidos: Custom order product with extended lead time. Contact your VWR Sales Representative for more information.