VWR® 18 mm Screw-Thread Headspace Vials and Caps
Supplier: VWR, part of Avantor
VWR®
10119-902A 10119-906A
10119-906APK 69.4 USD
10119-906A 10119-902A
VWR® 18 mm Screw-Thread Headspace Vials and Caps
Vials
Screw-thread magnetic headspace vials offer tool-free convenience of a screw-thread vial
  • Choose from all-steel or improved polymeric-metal version
  • Both compatible with all standard or magnetic Headspace autosamplers
  • Excellent sealing characteristics that withstand high pressures of headspace applications
Vials are converted from glass tubing to provide uniform wall thickness and promote even heat distribution.