VWR® Multiwell Cell Culture Plates
Supplier: VWR, part of Avantor
VWR®
TCP011024 TCP011196 TCP011096 TCP011048 TCP011001 TCP011384 TCP011006 TCP011004 TCP011012
10062-896CS 308.12 USD
10062-896 10062-902 10062-900 10062-898 75999-254 10814-226 10062-892 10814-222 10062-894
VWR® Multiwell Cell Culture Plates
Microplates Cell Culture Plates Coated Plates
VWR® Standard Multiwell Cell Culture Plates are plasma treated for consistent cell attachment and growth.
  • Clear polystyrene, sterile, certified non-pyrogenic
  • Well volume uniformity ensures uniform growth surface area exposure
  • Single-position lid reduces misplacement and the risk of contamination
  • Designed with alphanumeric code for easy well identification
Raised rims on wells and condensation rings on the lid reduce evaporation and minimize edge effects.
Certificaciones: ISO 9001, ISO 13485.
Información para pedidos: Individually wrapped in peel-to-open paper/plastic blister packs for all items except 75999-254, which is 10/pkg.