BRAND® nano-cap™ Pipette Tips, BrandTech®
Supplier: BrandTech
732022 732102 732002 732122 732222 732202
10033-996PK 263.81 USD
10033-996 10033-984 10034-028 10033-972 10034-126 10034-016
BRAND® nano-cap™ Pipette Tips, BrandTech®
Pipet Tips
BRAND nano-cap pipette tips are a shorter style (37 mm) ultra-micro/crystal tip for use on many pipettes with a nominal/maximum volume less than 20 µl.
  • Short (37 mm) ultramicro/"crystal" tips
  • Fits most major brands of pipettes
  • Suitable for Transferpette® sub-micro (0.1 - 1 µl) pipettes
All tips are produced under cleanroom conditions. Bulk tips are packed in resealable bags. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.
A unique capillary rise gives a visual check of pipetted volumes smaller than 1 microliter. Required for use with BRAND sub-micro (0.1 to 1 µl) Transferpette and Transferpette S models.
Certificaciones: CE-IVD.
Información para pedidos: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.