BRAND® Ultra Low Retention 50 µl Pipette Tips, BrandTech®
Supplier: BrandTech
732306 732326
10034-088PK 67.29 USD
10034-088 10034-046
BRAND® Ultra Low Retention 50 µl Pipette Tips, BrandTech®
Pipet Tips
BRAND Ultra-Low Retention pipette tips are produced through a patented physicochemical prodess that provides a homogeneous defect-free surface.
BRAND 50 µl pipette tips are a longer style (50 mm) ultra-micro/crystal tip for use on many pipettes with a nominal/maximum volume less than 20 microliters. Extended volume allows reverse pipetting of viscous or volatile liquids for greater accuracy. Graduations at 2.5, 10, 25 and 50 µl allow easy referencing of pipetted volume.
All tips are produced under cleanroom conditions. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.
This results in an extremely low surface tesnion, over 50% less than PTFE. This significantly reuces sample loss and improves reproducibility. BRAND Ultra Low Retention tips have no additives than can leach out, or coatings to wash off. Fully autoclavable at 121 °C without damage to surface properties.
Certificaciones: CE-IVD.
Información para pedidos: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.