BRAND® Pipette Tips, 300 µl, BrandTech®
Supplier: BrandTech
732130 732030 732230 732010 732110
10033-992PK 24.07 USD
10033-992 10033-980 10034-112 10034-134 10034-004
BRAND® Pipette Tips, 300 µl, BrandTech®
Pipet Tips
BRAND 300 µl pipette tips are a thin-walled low waste tip that fits virtually all pipettes with a yellow color code, as well as 300 µl multichannels.
  • Fits most major brands of 200/300 µl pipettes
  • Made under cleanroom conditions
  • All boxed/refill tips are DNA-/Rnase-/ATP-/endotoxin-free
All tips are produced under cleanroom conditions. Bulk tips are packed in resealable bags. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.
The additional volume makes this an excellent choice for reverse pipetting for greater accuracy with viscous or volatile liquids. Tip is 53mm long and has graduations at 50 and 100 µl to allow easy referencing of pipetted volume. All tips are colorless; racked tips are placed in a green colored tip tray for easy recognition.
Certificaciones: CE-IVD.
Información para pedidos: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.