Lab Coats, ESD Knit, White Knight
Supplier: White Knight
10032-314 10032-308 10032-770 10032-310 10032-312 10032-394 10032-768
10032-314EA 62.21 USD
10032-314 10032-768 10032-308 10032-394 10032-770 10032-310 10032-312
Lab Coats, ESD Knit, White Knight
Workwear
The ESD Knit Lab Coat is designed to provide excellent anti-static protection for electronics applications.
  • Excellent static dissipative properties
  • Lightweight, breathable fabric for added comfort when worn over regular clothes
  • Reinforced front facing and flat felled seams for improved durability
  • Three-position snap cuff for adjustability
The garments feature two lower pockets and one breast pocket. Each lab coat features a snap-front with reinforced facing and flat-felled seams for extra durability, and a three-position snap cuff.
Manufactured with a durable and breathable fabric, it offers a slight stretch for added comfort and ease of movement during asembly operations.
Ideal for use in micro-electronics assembly, microchip manufacturing, and semi-conductor production.
Información para pedidos: Available in blue only. Sizes 3X-Large and 4X-Large require extended lead time. Contact your VWR Sales Representative for more information.