VWR® Large Capacity Magnetic Stirrers
Supplier: VWR, part of Avantor
VWR®
80864027 80864026 80864020 80864021
12621-058EA 4875.19 USD
12621-058 10027-236 10027-090 12621-062
VWR® Large Capacity Magnetic Stirrers
Stirrers Magnetic Stirrers
Powerful magnetic drive system is capable of mixing high-viscosity materials.
  • Mix up to 200 L
  • Large stainless steel top plate
  • Built-in timer with alarm
  • Onboard memory recalls last set points
  • Now with Best-in-Class, Exclusive 5-Year Warranty
  • Available with a NIST Traceable, ISO/IEC 17025 Accredited Calibration Certificate from Troemner
Ideal for high volume applications up to 200L. Microprocessor, variable speed control provides consistent, uniform mixing action. Ramping feature slowly increases speed to setpoint for added safety and improved magnetic coupling. Independent LED display features speed and time. Timer will display elapsed time or when programmed to user-defined limit, will count down to zero and shut off unit. On-board memory will recall last speed and time settings, even when unit has been turned off. Audible alarm will sound when time reaches zero. Stirrer top plate is constructed of durable stainless steel and includes a removable nonskid mat.
Certificaciones: RoHS certified. TUV listed. Calibrated Stirrers include a Troemner ISO/IEC Accredited Calibration Certificate traceable to NIST. The ISO/IEC 17025 Accredited Calibration Certificate includes multiple data points within the speed range and time function with the associated uncertainties. Troemner is accredited by NVLAP under Laboratory Code 105013-0 to perform ISO/IEC 17025 calibrations.
Información para pedidos: All units include a 2.3m (92") detachable, three-wire electrical cord and plug. Model 1000 units are supplied with a 7 cm (23/4") egg shaped, PTFE-coated stir bar. Model 2000 units are supplied with a 10.2 cm (4") coated stir bar (with pivot ring). 230V, 50/60Hz models are also available; contact your VWR sales representative for more information.