Stripettor™ Ultra Pipet Controller, Corning®
Supplier: Corning
4099
10023-900CS 595.18 USD
10023-900
Stripettor™ Ultra Pipet Controller, Corning®
Pipets Pipet Controllers
This light, ergonomic, motorised pipette controller is designed for use with glass and plastic serological pipettes from 0.5 to 100 ml.
  • Conveniently positioned switches change operating modes and speeds quickly to handle different liquid volumes and various viscosities
  • Aspirating and dispensing speed is controlled through the finger triggers
  • When fully charged, the batteries enable up to 8 hours of continuous use
  • Large LCD display clearly indicates battery status, pipetting mode, and speed
  • Colored nose pieces for user or application identification
Información de suministro: Supplied with universal power supply, three 0.2 µm filters, 3 colored nose pieces and a wall hanger. Four samples of each Shorty Stripette serological pipette are also included.