VWR® Cryogenic Vials, Internally or Externally Threaded
Supplier: VWR, part of Avantor
VWR®
T301-1VW T308-5AVW T311-5VW T308-3AVW T309-1AVW T308-1AVW T308-5VW T311-4VW T309-5VW T309-4AVW T311-5AVW T308-2VW T311-1VW T310-5AVW T310-5VW T310-2VW T310-4AVW T308-2AVW T301-4VW T311-4AVW T301-4AVW T311-2VW T301-3VW T311-3VW T310-1AVW T309-3AVW T301-5AVW T309-2VW T301-5VW T310-2AVW T310-3AVW T310-10AVW T308-4AVW T301-2VW T309-2AVW T309-5AVW
10018-768CS 963.51 USD
10018-768 10018-746 10018-824 10018-748 10018-764 10018-742 10018-786 10018-820 10018-766 10018-788 10018-744 10018-822 10018-750 10018-772 10018-774 10018-730 10018-752 10018-770 10018-736 10018-758 10018-814 10018-738 10018-732 10018-776 10018-754 10018-734 10018-778 10018-756 10018-816 10018-818 10018-782 10018-760 10018-784 10018-762 10018-740 10018-780
VWR® Cryogenic Vials, Internally or Externally Threaded
Vials
Designed for storing biological material, human or animal cells at temperatures as low as –196 °C, but should be used only in the gas phase of liquid nitrogen.
  • Internal or external threaded closure options available
  • White marking area and printed graduations on all vials
  • Sterilized by gamma radiation
Closures and tubes are made of polypropylene with the same coefficient of expansion, which further enhances the leakproof qualities of these vials at various temperatures. Nonsterile polypropylene cap inserts provide color-coding for easy sample identification.
Each item has a large white marking area and printed graduations.
Please note that model 10018-740 has a polyethylene screw cap. Round bottom vials may be centrifuged up to 17000 g. Sterilized by gamma radiation (10⁻⁶), and autoclavable.
Certified RNase-, DNase-, Pyrogen- and DNA-free.
Tubes can be color coded by the use of CAPINSERT.
Certificaciones: CE marked.
Información para pedidos: Cap insert assortment includes 100 each of white, blue, red, green, and yellow.
Envase: Packaged in tamper-proof, resealable, safety-lock bags of 100.