VWR® Glassware Washers Accessories
Supplier: VWR, part of Avantor
437332 437271 437318 437158 437141 437196 437189 437233 437202 437219 437301 437134 437295 437226 437257 437127 437165 437172 437240 437264 437288 446508
10002-944EA 234.07 USD
10002-944 10002-934 10002-942 10002-910 10002-908 10002-918 10002-916 10002-926 10002-920 10002-922 10002-940 10002-906 10002-938 10002-924 10002-930 10002-904 10002-912 10002-914 10002-928 10002-932 10002-936 82021-092
VWR® Glassware Washers Accessories
Glassware Washers
Accessories designed for use with VWR® Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster pump for use with low pressure DI water supply below 18 psi
See details in table below for specific use information. For further assistance, view the product literature or contact your Avantor representative.