Would you like to log in again?
Tyvek® 800J Protective Garment, DuPont™
Supplier: DuPont
Tyvek® 800J is a chemical protective garment that combines resistance to low-concentration chemicals.
  • Water vapor permeable
  • Taped seams
  • Integrated chin flap and thumb loops
  • Good fit for hot, humid environments
  • Durable protection
Tyvek® 800J is a chemical-protective garment that combines resistance to low-concentration chemicals (even in pressurized form) with the comfort, protection, and durability of Tyvek®. Ideal applications for use include industrial and pressurized tank and machinery cleaning, lab animal research facilities, pharmaceutical API & chemical handling, food processing equipment maintenance, and waste treatment/disposal.
The garments' Tyvek® based technology is water vapor permeable, and features taped seams, an integrated chin flap and thumb loops. It is also a good fit for hot, humid environments, applications in which liquid and particulate protection is needed, and scenarios which require more protection than standard Tyvek® and less protection (and less basis weight) than Tychem®.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
We are currently unable to calculate your contract price for this item. List price is being displayed temporarily. When your order is processed, you will be invoiced at your contract price even though list price is displayed now
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
DuPont™ Tyvek® 800J Small TJ198TWHSM0025PI 10809-151 Case of 25 Retrieving
DuPont™ Tyvek® 800J Medium TJ198TWHMD0025PI 10809-131 Case of 25 Retrieving
DuPont™ Tyvek® 800J Large TJ198TWHLG0025PI 10809-133 Case of 25 Retrieving
DuPont™ Tyvek® 800J X-Large TJ198TWHXL0025PI 10809-135 Case of 25 Retrieving
DuPont™ Tyvek® 800J 2X-Large TJ198TWH2X0025PI 10809-137 Case of 25 Retrieving
DuPont™ Tyvek® 800J 3X-Large TJ198TWH3X0025PI 10809-155 Case of 25 Retrieving
DuPont™ Tyvek® 800J 4X-Large TJ198TWH4X0025PI 75854-084 Case of 25 Retrieving
DuPont™ Tyvek® 800J 5X-Large TJ198TWH5X0025PI 75854-086 Case of 25 Retrieving
DuPont™ Tyvek® 800J 6X-Large TJ198TWH6X0025PI 75854-088 Case of 25 Retrieving
DuPont™ Tyvek® 800J 7X-Large TJ198TWH7X0025PI 75854-090 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty