Thermo Scientific Pipette Service Support Plans
Whether it’s cGLP, cGMP, critical research, academia, or any special requirements you may need, our experienced service personnel are highly trained in all pipette models.
For more than 25 years, we have offered qualityminded service that meets our customers’ diverse needs. You will feel confident that your pipettes will have the accuracy and performance you require, when you need them.
Basic - Validation-Fast Check
The Validation-Fast Check Plan combines a thorough external pipette inspection with gravimetric validation to determine if desired accuracy and precision is achieved. This plan includes a validation pass/fail report. Ideal for customers who require:
  • An interim validation check that occurs in-between routine scheduled preventive maintenance and calibration
  • Repairs for dropped/disassembled pipettes or want to make sure pipettes are performing as desired
  • A cost-friendly service option
Quick, convenient and effective
Learn More
Preferred - Preventive Maintenance and Calibration
The Preventive Maintenance and Calibration Plan provides a thorough inspection, preventive maintenance and calibration. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
  • Pipette inspection, liquid leak test, routine preventive maintenance and minor repairs
  • Do not need as-received data
  • Calibration
Thorough, effective and dependable
Learn More
Premium - GxP Preventive Maintenance and Calibration
The GxP Preventive Maintenance and Calibration Plan provides a comprehensive service that includes gravimetric evaluation of your pipette’s performance prior to any inspection, preventive maintenance or minor repairs. This plan includes a calibration certificate and sticker.
Ideal for customers who require:
  • As-received data
  • Pipette inspection, liquid leak test, routine preventive maintenance and replacement of o-rings (as needed)
  • Calibration
  • Work in a highly regulated environment
  • Reduced OOTs (Out of Tolerance) that could invalidate research
Demanding, thorough and reliable
Learn More
Literature
Request literature or download PDFs from Thermo Scientific.