Automated Liquid Handlers and Dispensers
Increase productivity by reducing hands-on time with Thermo Scientific™ automated liquid handling and dispensing systems. These instruments handle a wide range of volumes and labware, providing fast, precise liquid handling for a variety of applications. Plus, their compact size and flexible programming options make them easy to integrate into your workflow.
Upgrade
Multidrop Reagent Dispensers
Thermo Scientific™ Multidrop™ dispensers provide fast, precise dispensing for your high-throughput lab. They handle a wide range of plate types and volumes, making them convenient solutions for a variety of applications.
Versette Automated Liquid Handler
Thermo Scientific™ Versette™ automated liquid handler provides an easy way to help minimize hands-on time required for liquid handling applications. This compact 96-/384-channel instrument is ideal for plate replication, plate stamping and serial dilution procedures.
Product videos
Multidrop Dispensers
Versette
Literature
Request literature or download PDFs from Thermo Scientific.
Promotions
View a comprehensive list of the latest Thermo Scientific promotions.