High-performance grip products with TracTek
Wet, Dry, or Oily TracTek® provides maximum grip in all conditions.
Feel the SW difference, compare TracTek styles to similar nitrile products for proof.
SW outperforms competitors without loss of sensitivity or durability.
The micro-diamond pattern channels liquid away from contact points for superior grip strength, protecting both the user and the product from slip-related accidents.
Product selection
POWERFORM® S8+ TracTek® nitrile gloves
POWERFORM® S8+ with TracTek delivers superior wet and oily grip. Rip-resistant, 2-ply construction provides tough ANSI Level-2 puncture protection.
HYPERSENSE® X9 TracTek® latex gloves
HYPERSENSE® X9 with TracTek delivers superior wet and oily grip. Pure latex construction provides unmatched tactile sensitivity, dexterity, and comfort.
VERTEX® X6 nitrile gloves
VERTEX® X6 with full-coverage grip. MilTek® optimizes thickness at wrist, palm, and fingers for rip resistance and dexterity.
POWERFORM® X5+ nitrile gloves
POWERFORM® X5+ with MilTek® optimizes thickness at wrist, palm, and fingers for rip resistance and dexterity.