Weight Sets
Precision-crafted of stainless steel, with a highly polished finish. Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking. Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.
Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove.