MasterSense™ Smart Peristaltic Pumps
Designed for today's critical process applications and the biopharma industry of tomorrow
Compare pump optionsIntuitive and innovative
- Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
- Store and recall standard protocols ensuring continuity
- Intuitive digital touchscreen with optimized menu navigation and program input, even with wet gloves on!
- The MasterSense drives offer 3 tiers of access, allowing for varying levels of programming all password protected, adhering to GMP best practices
Easy and accurate
- Operational modes include continuous run, volume dispense and time dispense
- Batch control and interval/off time in volume and time modes
- Qualitative tube calibration ensures tubing is calibrated precisely
- Graphical indicator lets you know when tubing is optimally calibrated
Connected and smart
- Options for connectivity and control include EtherNet/IP, Profibus, analog remote, and serial control
- MasterflexLive® allows you to monitor pump operation over the cloud -- via PC or mobile device
- Over-the-air firmware updates ensure that your MasterSense pump system has the latest features and functionality
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
Compare MasterSense Pumps
![]() |
Masterflex® B/T® MasterSense™ BioPharma Process Pump
|
highest flow 0.65 to 42 LPM |
pressure up to 35 psi |
Connectivity EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive |
![]() |
Masterflex I/P MasterSense™ Pumps
|
HIGH FLOW 0.001 to 26 LPM |
pressure up to 40 psi (2.7 bar) |
Connectivity EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive |
![]() |
Masterflex® L/S® MasterSense™ Pumps
|
medium flow 0.001 to 3400 mL/min |
pressure up to 150 psi (10.3 bar) |
Connectivity EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive |
![]() |
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex® Ultrapharm™ Pump Head
|
medium flow 0.0006 to 19 LPM |
pressure up to 150 psi (10.3 bar)? |
Connectivity EtherNet/IP or Profibus, plus MasterflexLive or analog remote control |
More Pumping Options With Advanced Touch-screen Operation
![]() |
Ismatec® Reglo Digital Multichannel Pumps
|
Low flow 0.0001 to 68 mL/min *depending on cartridge type |
pressure up to 35 psi |
Connectivity EtherNet/IP, analog remote, serial control, plus MasterflexLive |
![]() |
Ismatec Reglo Digital Miniflex® Pumps
|
Low flow 0.004 to 365 mL/min |
pressure up to 35 psi |
Connectivity EtherNet/IP, analog remote, serial control, plus MasterflexLive |
![]() |
Ismatec Reglo Digital Gear Pump Drive
|
Medium flow 1.7 to 5850 mL/min |
pressure up to 300 psi (20.7 bar) |
Connectivity EtherNet/IP, analog remote, serial control, plus MasterflexLive |
![]() |
Ismatec Reglo Digital Piston Pump Drive
|
Low flow 0.025 to 75 mL/min |
pressure up to 100 psi (6.9 bar) |
Connectivity EtherNet/IP, analog remote, serial control, plus MasterflexLive |
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs
Start now