HALO® Chromatography Columns and Consumables+-
HALO® columns are available in 10 phases for reversed-phase separations and two phases for hydrophilic interaction liquid chromatography (HILIC) separations. HALO® column packings are not made the typical way; instead, the particles packed into HALO® columns are manufactured using Fused-Core® particle technology that was developed to deliver hyper-fast liquid chromatographic separations. 
The original HALO® Fused-Core® columns are able to perform competitively versus sub-2 µm fully porous columns and substantially outperform 3 and 5 µm fully porous columns, delivering increased performance without the consequence of high back pressures. HALO® HPLC columns are designed to serve the pharmaceutical, biopharma, food and beverage, environmental, vitamin, industrial and clinical/toxicology market segments.