Constant Temperature Unit Validation, Calibration, and Installation
Avantor Services offers validation (IQ/OQ/PQ), installation, and calibration services for all makes, models, and configurations of constant temperature units, including (but not limited to) the following:
  • Ovens
  • Incubators
  • CO2 Incubators
  • Environmental Chambers
  • Water Baths
  • Circulators
  • Chillers
Please request a quote below. Or, feel free to get in touch with us at 1.888.793.2300 or equipmentservices.us@avantorsciences.com.
Request a Quote
Installation
Avantor Services will provide installation services as per your request. Our technicians ensure that your equipment is set up, powered on, and functioning properly, providing you with peace of mind.
Calibration & Tuning:
Calibrate and adjust the chamber using NIST traceable standards and ensure temperature stability. Calibration certificate provided.
IQ/OQ/PQ
Our qualification services will help you stay in compliance with FDA, GMP/GLP, US Pharmacopoeia, and ISO standards. Our IQ/OQ/PQ offers the following:
  • Installation Qualification: This will verify design, specifications, and system SOPs. IQ also verifies and documents environmental conditions, adequate space requirements, configuration, proper installation, spare parts lists/manuals, as well as SOPs provided by the customer.
  • Operation Qualification: The next step will test UI controls and functions, control panel, and other applicable monitors. We will then perform an empty unit temperature mapping for multiple hours to document the recovery time of the unit. Upon completion of temperature mapping, an open door and power loss temperature profile test will take place.
  • Performance Qualification: We re-perform temperature mapping at various points over multiple cycles with a loaded unit.             
Preventive Maintenance
Avantor Services offers a customized preventive maintenance program based on the specifications of your unit. Preventive maintenance helps to stop problems before they occur by preventing equipment failures and downtime. This service can maximize the life and performance of your equipment and instruments, while also helping the unit stay compliant by regulatory standards.